Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Alcohol Dehydrogenase from Yeast

Catalog No. p-4605309 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
15 kU
150 kU
30 kU
5 kU
75 kU
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10016105 5 kU
ICN10016115 15 kU
ICN10016175 75 kU
ICN10016130 30 kU
ICN10016181 150 kU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. ICN10016105 Supplier MP Biomedicals™ Supplier No. 0210016105
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Facilitates interconversion between alcohols and aldehydes or ketones with reduction of nicotinamide adenine dinucleotide

Alcohol dehydrogenases are a group of dehydrogenase enzymes that occur in many organisms. It is a metalloenzyme containing four tightly bound zinc atoms per molecule.Each subunit also contains a second zinc atom (conformational zinc), which stabilizes the enzyme's tertiary structure. Per subunit, there are two distinct active site sulfhydryl groups which can be distinguished on the basis of differential reactivity with iodoacetate and butyl isocyanate. A histidine residue is considered to have an essential role.

CAS Number: 9031-72-5

EC Number: 232-870-4

Format: Off-White Flaky Powder

Optimum pH: For the oxidation of ethanol, pH 8.6-9.0 (the enzyme becomes increasingly unstable at higher pH). For the reduction of acetaldehyde a pH nearer to 7.0 is considered optimum. This reaction is kinetically complex with pH being only one factor determining optimum conditions.

Solubility: Dissolves readily at 5 mg/ml in 0.01 M sodium phosphate pH 7.5 to give a clear colorless solution.

Activators: Sulfyhydryl activating reagents, mercaptoethanol, dithiothreitol, cysteine, etc., and heavy metal chelating reagents.

Stabilizers: Dilute solution of the enzyme may be stabilized by serum albumin, gelatin, and/or glutathione or cysteine. At pH values below 6.0 and above 8.5 the enzyme is increasingly unstable. More concentrated solutions of the enzyme in high purity water, near neutrality, are stable several days at 5oC.

In biotransformation, alcohol dehydrogenases are often used for synthesis of enantiomerically pure stereoisomers of chiral alcohols.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X
Color White
pH 8.6 to 9
For Use With (Application) Synthesis of enantiomerically pure Stereoisomers of chiral Alcohols, biotransformation
Content And Storage 4°C
Source Yeast
Quantity 5 kU
Product Type Alcohol Dehydrogenase
Molecular Weight (g/mol) 141000
Inhibitors Heavy Metals, -SH reagents
Form Lyophilized Powder
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.