Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Alcohol Oxidase

Catalog No. ICN19015501 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 kU
5 kU
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19015501 1 kU
ICN19015505 5 kU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19015501 Supplier MP Biomedicals™ Supplier No. 0219015501
Add to Cart
Add to Cart

From Pichea pastoris E.C.1.1.3.13

Catalyzes the oxidation of an alcohol to an aldehyde and hydrogen peroxide. Activity: 10-20U per mg protein. Unit Definition: One unit will oxidize 1.0μmole of methanol to formaldehyde per minute at pH 7.5 at 25°C. Solution in sucrose.

TRUSTED_SUSTAINABILITY

Specifications

Concentration ≥10 U/mg
Color Reddish Brown
pH 7.2
For Use With (Application) Colorimetric determination of Blood Alcohol
Storage Buffer 30% Sucrose, 0.1M Potassium Phosphate Buffer at pH 8
Content And Storage -20°C
Source Pichea pastoris
Quantity 1 kU
Product Type Alcohol Oxidase
Molecular Weight (g/mol) ≈630000
Form Frozen
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.