Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Aldehyde Dehydrogenase, Potassium Activated

Catalog No. p-3563326 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 U
250 U
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19067980 100 U
ICN19067983 250 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19067980 Supplier MP Biomedicals™ Supplier No. 0219067980
Add to Cart
Add to Cart

From yeast

Activity: 1U/mg solid (approx. 5U/mg protein). ADH Less than 0.1%. Unit Definition: One unit causes the oxidation of one micromole of acetaldehyde to acetic acid per minute at pH 8.0 and 25°C in the presence of potassium ions and thiols. Freeze dried stabilized powder.

TRUSTED_SUSTAINABILITY

Specifications

Concentration ≈5 U/mg Protein, ≥1 U/mg Solid
Color Off-White
pH 9
For Use With (Application) Oxidizes a number of Aliphatic and Aromatic Aldehydes
Content And Storage -20°C
Source Yeast
Quantity 100 U
Product Type Aldehyde Dehydrogenase
Molecular Weight (g/mol) ≈200000
Inhibitors SH-reagents, such as N-ethylmaleimide, 2-iodobenzoic Acid, Iodoacetic Acid and Chloromercuribenzoic Acid, Heavy Metals (especially Cu2-) and Acetaldehyde (at high concentrations)
Form Lyophilized Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.