Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Alkaline Phosphatase, Powder

Catalog No. ICN19068001 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 kU
10 kU
100 kU
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19068001 1 kU
ICN19068010 10 kU
ICN19068080 100 kU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN19068001 Supplier MP Biomedicals™ Supplier No. 0219068001
Add to Cart
Add to Cart

From calf intestine

Activity: ∽1.0U/mg. Unit Definition: One unit causes hydrolysis of one micromole of p-nitrophenyl phosphate per minute at pH 9.6 and 25°C. Freeze-dried powder.

TRUSTED_SUSTAINABILITY

Specifications

Concentration ≥1 U/mg
Color Off-White
pH 9.8
For Use With (Application) Used to remove 5'-phosphate groups from Nucleic Acid molecules to prevent self-ligation of DNA fragments and vectors during recombinant DNA manipulation
Content And Storage -20°C
Source Calf intestine
Quantity 1 kU
Product Type Alkaline Phosphatase
Molecular Weight (g/mol) 140000
Inhibitors Acidification to 4.5 pH reversibly inactives the Enzyme, Inorganic Phosphate, chelating agents, EDTA
Form Lyophilized Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.