Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ b-Lactamase [E.C. 3.5.2.6]

Catalog No. ICN19003401 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 Vial
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19003401 1 Vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19003401 Supplier MP Biomedicals™ Supplier No. 0219003401
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

From Bacillus cereus

Lyophilized solid. Activity: Each vial contains 500U β-Lactamase I and 50U β-Lactamase II. Unit Definition: One unit is defined as the amount of enzyme which will hydrolyze 1.0μm of benzyl penicillin and 1.0μm of cephalosporin C per minute at 25°C in the presence of EDTA pH 7.0.

TRUSTED_SUSTAINABILITY

Specifications

pH 7
For Use With (Application) Destroy beta-Lactam antibiotics in body fluids or culture media and It is used in the production of Penicillin
Content And Storage 4°C
Source Bacillus cereus
Quantity 1 Vial
Product Type b-Lactamase
Molecular Weight (g/mol) 32000
Inhibitors Clavulanic Acid, Sulbactam, Tazobactam
Form Lyophilized Solid
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.