Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ beta-Glucosidase from Sweet Almonds

Catalog No. ICN10034890 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 kU
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10034890 500 kU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10034890 Supplier MP Biomedicals™ Supplier No. 0210034890

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

Involved in hydrolysis of β-glycosidic bonds connecting carbohydrate residues in β-D-glycosides

  • Synonyms: β-D-Glucoside glucohydrolase
  • CAS Number: 9001-22-3
  • EC Number: 232-589-7
  • MDL Number: MFCD00130628
  • Format: Lyophilized powder, salt free
  • Specific Activity: >2500U/mg solid
  • Dissolves readily at 5mg/mL in 0.1M sodium acetate (pH 5.0) to give clear pale yellow solution

  • Involved in synthesis of glucosides and fucosides
  • Used in pharmaceutical, cosmetic and detergent industries
  • Hydrolytic removal of aglycone moiety from flavonoid and isoflavonoid glycosides
  • Flavor enhancement of fruit juices and wine
  • Biosynthesis of oligosaccharides

TRUSTED_SUSTAINABILITY

Specifications

Concentration >2500 U/mg
Color Pale Yellow
For Use With (Application) Potential applications in pharmaceutical, cosmetic and detergent industries, hydrolytic removal of aglycone moiety from Flavonoid and Isoflavonoid Glycosides, flavor enhancement of fruit juices and wine and biosynthesis of Oligosaccharides
Content And Storage -20°C
Source Sweet almonds
Quantity 500 kU
Product Type beta-Glucosidase
Form Lyophilized Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.