Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Beta-Glucuronidase from Helix pomatia

Catalog No. ICN15228405 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15228405 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15228405 Supplier MP Biomedicals™ Supplier No. 0215228405
Add to Cart
Add to Cart

Routinely used for the enzymatic hydrolysis of glucuronides from Serum plasma and other fluids prior to analysis by enzyme immunoassay, mass spectrometry, gas chromatography, high performance liquid chromatography, or other means.

TRUSTED_SUSTAINABILITY

Specifications

Concentration >50,000 U/mL
Color Brown
pH 5
For Use With (Application) Used as reporter gene enzymatic hydrolysis of Glucuronides
Content And Storage 2°C to 8°C
Source Helix pomatia
Quantity 5 mL
Product Type beta-Glucuronidase
Inhibitors D-glucuronic Acid, D-galacturonic Acid and D-glucaro-1,4-lactone
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.