Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Bilirubin Oxidase

Catalog No. ICN19000080 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 U
100 U
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19000080 100 U
ICN19000010 10 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19000080 Supplier MP Biomedicals™ Supplier No. 0219000080
Add to Cart
Add to Cart

From Myrothecium species

Activity: ≥0.8U/mg solid. Unit Definition: One unit is the amount of enzyme that oxidizes one micromole of bilirubin per minute at 37°C, pH 8.4. Lyophilized powder.

TRUSTED_SUSTAINABILITY

Specifications

Concentration ≥0.8 U/mg
Color Light Blue/Green
For Use With (Application) Degrade Bilirubin, used to determine free hemoglobin in icteric specimens, also has potential application in dye effluent decolorization and is a potential treatment for neonatal jaundice
Content And Storage -20°C
Source Myrothecium species
Quantity 100 U
Product Type Bilirubin Oxidase
Molecular Weight (g/mol) 52000
Form Lyophilized Powder
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.