Learn More
MP Biomedicals™ Bovine Albumin, from New Zealand-sourced Bovine Plasma
Shop All MP Biomedicals, Inc ProductsDescription
Certain conformational and primary-sequence epitopes of BSA are suspected allergens in human beef and milk allergies
- From New Zealand-sourced Bovine Plasma
- Microbiological Grade
- ≥97% Total Protein
- Yellow to Brown Powder
- Synonyms: BSA, Bovine albumin, Albumin from bovine serum
- CAS Number: 9048-46-8
- EC Number: 232-936-2
- MDL Number: MFCD00130384
Specifications
Specifications
| Formulation | ≥97% Total Protein |
| Name | Bovine Albumin |
| Quantity | 500 g |
| Source | Bovine |
| Form | Powder |
| Purity or Quality Grade | Microbiological Grade |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.