Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Bovine Albumin, from New Zealand-sourced Bovine Plasma

Catalog No. MP218062091 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
100 g
500 g
1 kg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP218062091 1 kg
MP218062025 25 g
MP218062010 10 g
MP218062080 100 g
MP218062090 500 g
MP218062050 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. MP218062091 Supplier MP Biomedicals™ Supplier No. 0218062091
Add to Cart
Add to Cart
Request a Quote

Certain conformational and primary-sequence epitopes of BSA are suspected allergens in human beef and milk allergies

  • From New Zealand-sourced Bovine Plasma
  • Microbiological Grade
  • ≥97% Total Protein
  • Yellow to Brown Powder
  • Synonyms: BSA, Bovine albumin, Albumin from bovine serum
  • CAS Number: 9048-46-8
  • EC Number: 232-936-2
  • MDL Number: MFCD00130384

TRUSTED_SUSTAINABILITY

Specifications

Formulation ≥97% Total Protein
Name Bovine Albumin
Quantity 1 kg
Source from New Zealand-sourced Bovine Plasma
Form Powder
Purity or Quality Grade Microbiological Grade
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.