Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Bovine Albumin, Technical Grade

Catalog No. MP218069750 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
50 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP218069750 50 g
MP218069780 100 g
MP218069725 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP218069750 Supplier MP Biomedicals™ Supplier No. 0218069750

  • From New Zealand-sourced Bovine Plasma
  • Technical Grade

TRUSTED_SUSTAINABILITY

Specifications

Name Bovine Albumin
Quantity 50 g
Source Bovine
Purity or Quality Grade Technical Grade
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.