Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Bovine Transferrin, (HOLO)

Catalog No. ICN15233590 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15233590 500 mg
ICN15233580 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15233590 Supplier MP Biomedicals™ Supplier No. 0215233590
Add to Cart
Add to Cart

Suitable for regulating iron levels in cell culture

Transferrin is a 77kDa plasma glycoprotein with the capacity to bind ferric ions and as such is a source of bio-available iron. The product is frequently used as a nutrient in cell and microbial culture and as a media supplement in the production of bio-pharmaceutical products. Transferrin has also been shown to function as a factor in the growth stimulation of many cell types, such as granulocytes, macrophages and endothelial cells. Chromatographically purified.

Physical characteristics:

  • Appearance: Reddish-brown powder
  • Purity: ≥95% w/w
  • Iron: ≥1200 mg/g protein
  • Moisture: ≤5%
  • pH (3%): 6.0 to 8.0
  • Solubility: Dissolves in <20 min.
Microbial evlauation and control:
  • Bioburden: ≤100cfu/g
  • Endotoxin: ≤1 EU/mg
  • Mycoplasma: Not detected
  • Virus: Not detected
Certifications: ISO9000, cGMP, EDQM

TRUSTED_SUSTAINABILITY

Specifications

pH Range 6 to 8
Quantity 500 mg
Source Bovine Serum
Storage Requirements 4°C
Product Type Bovine Transferrin (HOLO)
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.