Learn More
Description
Casein hydrolysate enzymatic is recommended for use as an animal diet.
Specifications
Specifications
| Product Type | Animal Diet |
| Components | Moisture 3.5%, Amino N/Total N 50.3%, Chloride 0.8%, Iron <50 ppm, Solubility 25 g/L, Total Nitro gen 12.8%, Sodium 2.8%, Calcium 0.1%, pH (2% solution) 6.6, Ammonia 0.9 |
| Format | Powder |
| For Use With (Application) | Casein Hydrolysate Enzymatic is recommended for use as animal diet, also recommended for the production of media for the growth of Charon phage and other lambda phages |
| Quantity | 454 g |
| Storage Requirements | 15°C to 30°C |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.