Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Casein Hydrolysate Enzymatic, Soluble, Powder

Soluble; NZ Amine A ; Peptone from casein

$290.50 - $991.50

Specifications

Product Type Animal Diet
Components Moisture 3.5%, Amino N/Total N 50.3%, Chloride 0.8%, Iron <50 ppm, Solubility 25 g/L, Total Nitro gen 12.8%, Sodium 2.8%, Calcium 0.1%, pH (2% solution) 6.6, Ammonia 0.9
Format Powder
For Use With (Application) Casein Hydrolysate Enzymatic is recommended for use as animal diet, also recommended for the production of media for the growth of Charon phage and other lambda phages
Storage Requirements 15°C to 30°C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ICN96013801
View Documents
MP Biomedicals™
0296013801
454 g
Each for $290.50
Only null left
Add to Cart
 
ICN96013805
View Documents
MP Biomedicals™
0296013805
5 lb.
Each for $991.50
Only null left
Add to Cart
 
Description

Description

Casein hydrolysate enzymatic is recommended for use as an animal diet.

Specifications

Specifications

Animal Diet
Powder
15°C to 30°C
Moisture 3.5%, Amino N/Total N 50.3%, Chloride 0.8%, Iron <50 ppm, Solubility 25 g/L, Total Nitro gen 12.8%, Sodium 2.8%, Calcium 0.1%, pH (2% solution) 6.6, Ammonia 0.9
Casein Hydrolysate Enzymatic is recommended for use as animal diet, also recommended for the production of media for the growth of Charon phage and other lambda phages
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.