Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Catalase

Catalog No. ICN10040250 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10040250 50 mL
ICN10040280 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10040250 Supplier MP Biomedicals™ Supplier No. 0210040250
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fungal suspension from Aspergillus niger

Catalase, [9001-05-2]

Density: 1.2g/mL
Activity: 1000 units/ml

TRUSTED_SUSTAINABILITY

Specifications

Concentration ≥1000 U/mL
For Use With (Application) Removal of Peroxides, the generation of Oxygen and in coupled systems, for the determination of metabolites
Content And Storage 4°C
Source Aspergillus niger
Quantity 50 mL
Product Type Catalase
Molecular Weight (g/mol) ≈250
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.