Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Catalase from Bovine Liver

Catalog No. icn10042910 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10042910 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10042910 Supplier MP Biomedicals™ Supplier No. 0210042910
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Natural antioxidant used to study roles of reactive oxygen species in gene expression and apoptosis

A homotetrameric enzyme that is primarily located in peroxisomes. It is a tetramer consisting of 4 equal subunits with a molecular weight of 60kDa each. Each subunit contains iron bound to a protoheme IX group. The enzyme also strongly binds NADP, which is in close proximity to the heme group (13.7Å apart). Catalase can also react with alkylhydrogen peroxides such as methylperoxide and ethylperoxide and does not require any activator for exhibiting its catalytic activity. Catalase activity is constant over the pH range of 4.0 to 8.5.

 

  • CAS Number: 9001-05-2
  • EC Number: 232-577-1
  • MDL Number: MFCD00081483
  • Format: Clear, pale yellow solution
  • Specific Activity: ∽30,000U/mL; ≥40,000U/mg protein
  • Extinction Coefficient: 36.5 (276nm)
  • Sterile Aqueous Solution, miscible in water

 

TRUSTED_SUSTAINABILITY

Specifications

Concentration ≈30,000 U/mL, ≈40,000 U/mg Protein
Color Pale Yellow
pH 4 to 8.5
For Use With (Application) Used as an antioxidant in cell culture media, used to study the role of ROSs in gene expression and apoptosis, mainly in cancer researches and it is also used to eliminate H2O2 from enzymatic Oxidation reactions
CAS 9001-05-2
Content And Storage 4°C
Sterility Sterile Aqueous Solution
Source Bovine liver
Quantity 10 mL
Product Type Catalase
Molecular Weight (g/mol) 250
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.