Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Catalase from Aspergillus niger (E.C.1.11.1.6)

Catalog No. ICN19031101 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mU
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19031101 1 mU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19031101 Supplier MP Biomedicals™ Supplier No. 0219031101
Add to Cart
Add to Cart

Lyophilized powder

Activity: 2000U/mg material. Unit Definition: One unit decomposes oneμmole of hydrogen peroxide per minute at 25°C, pH 7.0.

Ref.: Academic Press, New York, Based on the method of Bergmeyer, Methods of Enzymatic Analysis, 438, (1974).

TRUSTED_SUSTAINABILITY

Specifications

Concentration 5 mg/mL
Color Brown to Tan
For Use With (Application) Removal of Peroxides and generation of Oxygen
Content And Storage -20°C
Source Aspergillus niger
Quantity 1 mU
Product Type Catalase
Molecular Weight (g/mol) ≈240000
Form Lyophilized Solid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.