Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ CellMaxx™ Bovine Albumin, Low Endotoxin, Stem Cell Culture

Catalog No. MP219989625 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
100 g
500 g
1000 g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219989625 25 g
MP219989610 10 g
ICN989650 50 g
MP219989690 500 g
MP219989691 1000 g
MP219989680 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. MP219989625 Supplier MP Biomedicals™ Supplier No. 0219989625
Add to Cart
Add to Cart

Lipid-rich bovine serum albumin, suitable for stem cell culture and media formulation

Chromatographically purified for culturing embryonic, mesenchymal and hematopoietic cells. Lipid-rich formulation can be substituted for Albumax™ II.

Physical characteristics:

  • Form: Off white to brown powder
  • Purity:≥98% (w/w)
  • Total Protein: ≥96%
  • pH (4%): 6.5 to 7.5
  • Solubility: Dissolves in 20 minutes at 20° to 25°C
  • Moisture: ≤5% w/w
Microbial evaluation and control:
  • Endotoxin: ≤1EU/mg
  • Mycoplasma: Negative
  • Bioburden: ≤100cfu/g
  • Virus (9CFR): Negative
  • Reported: Lipid content

Certifications: ISO9001, ISO13485, cGMP, EDQM

 

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Mammalian Cell Culture
Formulation Lipid-rich
Molecular Weight (g/mol) 66
Name CellMaxx™ Bovine Albumin, Low Endotoxin
pH Range 6.5 to 7.5
Purification Method Chromatography
Quantity 25 g
Source Bovine
Storage Requirements 4°C
Form Powder
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.