Learn More
MP Biomedicals™ CellMaxx™ Bovine Albumin, Low Endotoxin, Stem Cell Culture
Shop All MP Biomedicals, Inc ProductsDescription
Chromatographically purified for culturing embryonic, mesenchymal and hematopoietic cells. Lipid-rich formulation can be substituted for Albumax™ II.
Physical characteristics:
- Form: Off white to brown powder
- Purity:≥98% (w/w)
- Total Protein: ≥96%
- pH (4%): 6.5 to 7.5
- Solubility: Dissolves in 20 minutes at 20° to 25°C
- Moisture: ≤5% w/w
- Endotoxin: ≤1EU/mg
- Mycoplasma: Negative
- Bioburden: ≤100cfu/g
- Virus (9CFR): Negative
- Reported: Lipid content
Certifications: ISO9001, ISO13485, cGMP, EDQM
Specifications
Specifications
| For Use With (Application) | Mammalian Cell Culture |
| Formulation | Lipid-rich |
| Molecular Weight (g/mol) | 66 |
| Name | CellMaxx™ Bovine Albumin, Low Endotoxin |
| pH Range | 6.5 to 7.5 |
| Purification Method | Chromatography |
| Quantity | 100 g |
| Source | Bovine |
| Storage Requirements | 4°C |
| Form | Powder |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.