Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ CellMaxx™ Bovine Albumin, Low Free Fatty Acid, Stem Cell Culture

Catalog No. MP219989991 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
100 g
500 g
1000 g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219989991 1000 g
MP219989910 10 g
MP219989925 25 g
ICN989950 50 g
MP219989980 100 g
MP219989990 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. MP219989991 Supplier MP Biomedicals™ Supplier No. 0219989991
Add to Cart
Add to Cart

Bovine serum albumin suitable for stem cell culture and media formulation requiring very low levels of lipids

Chromatographically purified for culturing embryonic, mesenchymal and hematopoietic cells. Features low endotoxin, low level of lipids, fatty acids and hormones, e.g. insulin, T3, T4.

Physical characteristics:

  • Form: Off white to brown powder
  • Purity: ≥98% (w/w)
  • Total Protein: ≥96%
  • Free Fatty Acids ≤0.005% w/w
  • pH (4%): 6.5 to 7.5
  • Solubility: Dissolves in 20 minutes at 20° to 25°C
  • Moisture: ≤5% w/w
Microbial evaluation and control:
  • Endotoxin: ≤1EU/mg
  • Mycoplasma: Negative
  • Bioburden: ≤100cfu/g
  • Virus (9CFR): Negative
  • Reported: Insulin, T3, T4
Certifications: ISO9000, cGMP, EDQM

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Mammalian Cell Culture
Molecular Weight (g/mol) 66
Name CellMaxx™ Bovine Albumin
pH Range 6.5 to 7.5
Purification Method Chromatography
Quantity 1000 g
Source Bovine
Storage Requirements 4°C
Form Powder
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.