Learn More
MP Biomedicals™ CellMaxx™ Bovine Albumin, Low IgG, Stem Cell Culture
Shop All MP Biomedicals, Inc ProductsDescription
Chromatographically purified for culturing embryonic, mesenchymal and hematopoietic cells; IgG removed to very low levels. Lipid-rich formulation can be substituted for Albumax I™.
Physical characteristics:
- Form: Off white to brown powder
- Purity: ≥98% (w/w)
- Total Protein: ≥96%
- IgG 0.005% w/w
- pH (4%): 6.5 to 7.5
- Solubility: Dissolves in 20 minutes at 20° to 25°C
- Moisture: ≤5% w/w
- Endotoxin: ≤1EU/mg
- Mycoplasma: Negative
- Bioburden: ≤100cfu/g
- Virus (9CFR): Negative
- Reported: Lipid content
Specifications
Specifications
| For Use With (Application) | ELISA, Mammalian Cell Culture, Western Blot |
| Formulation | Lipid-rich |
| Molecular Weight (g/mol) | 66 |
| Name | CellMaxx™ Bovine Albumin, Low IgG |
| pH Range | 6.5 to 7.5 |
| Purification Method | Chromatography |
| Quantity | 50 g |
| Source | Bovine |
| Storage Requirements | 4°C |
| Form | Powder |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.