Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Cellulase from Aspergillus niger

Catalog No. ICN15058305 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15058305 5 g
ICN15058325 25 g
ICN15058380 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15058305 Supplier MP Biomedicals™ Supplier No. 0215058305
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cellulase from Aspergillus niger catalyzes hydrolysis of endo-1,4-β-D-glycosidic linkages in cellulose, lichenin, barley glucan and cellooligosaccharides cellotriose to cellohexaose

Cellulase refers to a group of enzymes which acting together hydrolyze cellulose. The b-glucosidases, including cellobiose are primarily active on the smaller molecular weight cellulose hydrolysates.

  • CAS Number: 9012-54-8
  • EC Number: 232-734-4
  • MDL Number: MFCD00081510
  • Format: Tan powder
  • Specific activity: >60000U/g
  • Readily soluble in water
  • Foreign activity: Salmonella Negative
  • Optimum pH: 4.2 to 5.2

Cellulase is used in molecular biology, cell biology, textile industry, pulp and paper industry and for commercial food processing in coffee

TRUSTED_SUSTAINABILITY

Specifications

Concentration >60,000 U/g
Color Tan
pH 4.2 to 5.2
For Use With (Application) Cellulase is used for commercial food processing in coffee, textile industry and in laundry detergents, pharmaceutical applications, fermentation of biomass into biofuels
Content And Storage 4°C
Source Aspergillis niger
Quantity 5 g
Product Type Cellulase
Form Powder
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.