Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Choline Oxidase from Alcaligones Species

Catalog No. ICN15349291 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 U
1000 U
500 U
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15349291 1000 U
ICN15349280 100 U
ICN15349290 500 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15349291 Supplier MP Biomedicals™ Supplier No. 0215349291
Add to Cart
Add to Cart

>10 units/mg solid, Yellowish powder. For cholinesterase activity assays.

TRUSTED_SUSTAINABILITY

Specifications

Concentration >10 U/mg
Color Yellow
pH 8 to 8.5
For Use With (Application) Enzymatic determination of Phospholipids by coupling with Phospholipase D and for Cholinesterase activity assays
Content And Storage -20°C
Source Alcaligenes species
Quantity 1000 U
Product Type Choline Oxidase
Molecular Weight (g/mol) ≈95000
Inhibitors p-Chloromercuribenzoate, Cu++, Co++, Hg++, Ag+
Form Lyophilized Powder
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.