Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Choline Oxidase

Catalog No. ICN15349250 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15349250 50 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15349250 Supplier MP Biomedicals™ Supplier No. 0215349250

missing translation for 'validMainframeMsg'

From Alcaligones sp.

Choline Oxidase, [9028-67-5]

  • Lyophilized powder
  • Activity: 10 units/mg solid or more (containing approx. 20% of stabilizers)

TRUSTED_SUSTAINABILITY

Specifications

Concentration >10 U/mg
Color Yellow
For Use With (Application) Enzymatic determination of Phospholipids
Storage Buffer Tris HCl Buffer 10mM, pH 8 containing EDTA 2mM and KCl 1%
Content And Storage -20°C
Source Alcaligenes species
Quantity 50 U
Product Type Choline Oxidase
Molecular Weight (g/mol) ≈95000
Inhibitors p-Chloromercuribenzoate, Cu++, Co++, Hg++, Ag+
Form Lyophilized Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.