Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ CIRCLEGROW™ Large Capsules

Catalog No. MP113000126 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
454 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP113000126 454 g
MP113000136 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP113000126 Supplier MP Biomedicals™ Supplier No. 113000126
Add to Cart
Add to Cart

Perfect for overnight or weekend cultures, CIRCLEGROW Medium is a super-rich bacterial growth medium that produces up to three times more plasmid DNA than traditional growth medium

CIRCLEGROW Medium consists of a protein and amino acid source, biological buffer, and a complex carbon source that gets metabolized at a much slower rate than glucose, thus prolonging the cell cycle and preventing cell death
during the log phase of growth. Cells grown with CIRCLEGROW Medium remain healthy for more than two days and continue to divide slowly after the log phase of the growth has been completed.

  • Reduce hands-on work and preparation time
  • Cultures do not need to be constantly monitored by O.D. readings due to the incremental post-log cell growth
  • Obtain excellent yields of plasmid DNA from smaller culture volumes
  • Confidently leave cultures overnight or over the weekend
  • Capsules are the most convenient form of this unique growth media
  • Pre-mixed CIRCLEGROW is also available in bulk powder, single-use pouches and sterile liquid
Formulation
  • Use 8 large capsules per liter;
  • Contents/liter: Proprietary Formulation

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Molecular biology Certified™
Content And Storage 15°C to 30°C
Form Dehydrated
Grade Molecular biology
Product Type Microbiology Media
Formulation Yeast extract, Tryptone broth, Vitamins, Biological Buffers have been added for proper pH adjustment
For Use With (Application) For growth of Escherichia coli, plasmid and certified for plasmid and other bacterial cultures in liquid medium
Quantity 454 g
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.