Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ CIRCLEGROW™, Powder

Catalog No. MP113000132 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 x 0.5 L
10 x 1 L
2.3 kg
227 g
454 g
5 kg
908 g
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP113000132 908 g
MP113000122 454 g
MP113000165 10 x 0.5 L
MP113000175 10 x 1 L
MP113000152 5 kg
MP113000112 227 g
MP113000142 2.3 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. MP113000132 Supplier MP Biomedicals™ Supplier No. 113000132
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Superior plasmid yield through enhanced cell growth

Perfect for overnight or weekend cultures, CIRCLEGROW Medium is a super-rich bacterial growth medium that produces up to three times more plasmid DNA than traditional growth medium. CIRCLEGROW Medium consists of a protein and amino acid source, biological buffer, and a complex carbon source that gets metabolized at a much slower rate than glucose, thus prolonging the cell cycle and preventing cell death during the log phase of growth.

  • Cells grown with CIRCLEGROW Medium remain healthy for more than two days and continue to divide slowly after the log phase of the growth has been completed
  • Reduce hands-on work and preparation time
  • Cultures do not need to be constantly monitored by O.D. readings due to the incremental post-log cell growth
  • Obtain high yields of plasmid DNA from smaller culture volumes, and confidently leave cultures overnight or over the weekend
  • Capsules are the most convenient form of this unique growth media
  • Pre-mixed CIRCLEGROW is also available in single-use pouches, capsules and sterile liquid
Formulation
  • MP113000165: Includes ten 20 g pouches. Add contents of one pouch to 500mL of purified water.
  • MP113000175: Includes ten 40g pouches. Add contents of one pouch to 1L of purified water.
  • MP113000185: Includes one hundred 20g pouches. Add contents of one pouch to 500mL of purified water.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15° to 30°C
Product Type Bacterial Growth Medium
Sterility Sterile
pH 6.9
Quantity 908 g
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.