Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Cod Liver Oil

Catalog No. ICN90140501 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 qt.
4 qt.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN90140501 1 qt.
ICN90140504 4 qt.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN90140501 Supplier MP Biomedicals™ Supplier No. 0290140501
Add to Cart
Add to Cart

Cod liver oil is a nutritional supplement derived from liver of cod fish, it has high levels of the omega-3 fatty acids, EPA and DHA.

Cod liver oil is a nutritional supplement derived from liver of cod fish, it has high levels of the omega-3 fatty acids, EPA and DHA.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Animal Diet
Components Vitamin A: min 850 USP μ/g, Vitamin D: min 85 USP μ/g
For Use With (Application) Cod liver Oil is a source of Omega-3 Fatty Acid
Quantity 1 qt.
Storage Requirements 15°C to 30°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.