Learn More
Description
- CAS Number: 9007-34-5
- EC Number: 232-697-4
- MDL Number: MFCD00130825
- Format: Powder
- Soluble in 0.075M sodium citrate pH 3.7 and in dilute acetic acid
For attachment of primary cultures of epithelioid cells and many other cell types
Specifications
Specifications
| For Use With (Application) | Tumor Marker |
| pH Range | 3.7 |
| Quantity | 100mg |
| Source | Calf Skin |
| Product Type | Chorionic Gonadotropin from Human Pregnancy Urine |
| Content And Storage | -20°C |
| Form | Lyophilized |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.