Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Collagenase, From Clostridium histolyticum

Catalog No. ICN15070501 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15070501 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15070501 Supplier MP Biomedicals™ Supplier No. 0215070501
Add to Cart
Add to Cart

Suitable for isolation of cells from many types of animal tissue.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 mg/mL
Color Brown
pH 7 to 9
For Use With (Application) Cell dissociation and cell lysis
Storage Buffer 1mg/mL of Enzyme in 0.05M TES with 0.36mM Calcium Chloride, pH 7.5
Content And Storage 4°C
Source Clostridium histolyticum
Quantity 1 g
Product Type Collagenase
Molecular Weight (g/mol) 105000 and 57400
Inhibitors Metal chelating agents such as Cysteine, EDTA or o-Phenanthroline
Form Lyophilized Solid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.