Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Collagenase from Clostridium histolyticum

Catalog No. ICN1951093 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN1951093 25 mg
ICN1951091 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN1951093 Supplier MP Biomedicals™ Supplier No. 02195109.3
Add to Cart
Add to Cart

Enzyme responsible for break down of peptide bonds in collagen.

Assist in destroying extracellular structures in pathogenesis of bacteria such as Clostridium. These enzymes are exotoxins (a virulence factor) and help to facilitate the spread of gas gangrene. They normally target the connective tissue in muscle cells and other body organs. 

  • Synonyms: Clostridiopeptidase A
  • CAS Number: 9001-12-1
  • EC Number: 232-582-9
  • MDL Number: MFCD00130830
  • Format: Dialyzed, lyophilized brown lumpy powder
  • Specific Activity: >125U/mg
  • Solubility: Dissolve enzyme at concentration of 1mg/mL in 0.05M TES with 0.36mM calcium chloride, pH 7.5 

Note: This item is for research use only.  It is not a compendial item and is not meant for medical use.

TRUSTED_SUSTAINABILITY

Specifications

Concentration >125 U/mg
Color Brown
pH 7 to 9
Content And Storage 4°C
Source Clostridium histolyticum
Quantity 25 mg
Product Type Collagenase
Molecular Weight (g/mol) 105000 and 57400
Inhibitors Metal chelating agents such as Cysteine, EDTA or o-Phenanthroline
Form Lyophilized Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.