Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ CSM-HIS-TRP, Powdered Media

Catalog No. MP114520112 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MP114520112 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP114520112 Supplier MP Biomedicals™ Supplier No. 114520112
Add to Cart
Add to Cart

Dropout of Complete Supplement Mixture (CSM) of amino acids for ;S. cereviseae growth media

Formulation

  • Add 0.72g of CSM-His-Trp to each liter of DOB or DOBA and autoclave at 121°C for 25 minutes for complete synthetic defined double drop-out medium

Applications:

Transcription regulation, Gene expression

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Molecular biology Certified™
Content And Storage 15°C to 30°C
Form Powder
Product Type Yeast Media
Formulation Adenine 10mg/L, L-Arginine HCl 50mg/L, L-Aspartic Acid 80mg/L, L-Histidine HCl 0mg/L, L-Isoleucine 50mg/L, L-Leucine 100mg/L, L-Lysine HCl 50mg/L, L-Methionine 20mg/L, L-Phenylalanine 50mg/L, L-Threonine 100mg/L, L-Tryptophan 0mg/L, L-Tyrosine 50mg/L, Uracil 20mg/L, L-Valine 140mg/L
For Use With (Application) For growth of Saccharomyces cerevisiae
Quantity 10 g
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.