Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Deoxyribonuclease I from Bovine Pancreas

Catalog No. ICN10057505 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 mg
20 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10057505 5 mg
ICN10057520 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10057505 Supplier MP Biomedicals™ Supplier No. 0210057505
Add to Cart
Add to Cart

Used as genetic marker for forensic identification

An endonuclease that splits phosphodiester linkages, preferentially adjacent to a pyrimidine nucleotide. The average chain of limit digest is a tetranucleotide. DNase I acts upon single chain DNA, and upon double-stranded DNA and chromatin.

  • CAS Number: 9003-98-9
  • EC Number: 232-667-0
  • MDL Number: MFCD00130918
  • Format: White powder

TRUSTED_SUSTAINABILITY

Specifications

Concentration ≥2000 kU/mg
Color White
pH 7.8
For Use With (Application) Used as a genetic marker for forensic identification
Content And Storage 4°C
Source Bovine pancreas
Quantity 5 mg
Product Type Deoxyribonuclease I
Molecular Weight (g/mol) 31000
Inhibitors EDTA 7,8 and Sodium Dodecyl Sulfate
Form Lyophilized Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.