Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Diaphorase from Clostridium kluyveri

Catalog No. ICN15084301 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1.8 kU
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15084301 1.8 kU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15084301 Supplier MP Biomedicals™ Supplier No. 0215084301
Add to Cart
Add to Cart

Used to assess the protein-protein interactions in assembly of lipoic acid on the 2-oxoacid dehydrogenases of aerobic metabolism

TRUSTED_SUSTAINABILITY

Specifications

Color Dark Golden
Content And Storage 0°C
Quantity 1.8 kU
Product Type Diaphorase, Lipoamide Dehydrogenase, Lipoyl Dehydrogenase
Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.