Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Diaphorase from Porcine Heart

Catalog No. ICN15084405 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
Concentration:
≈40 U/mg
25 U/mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
ICN15084405 5 mg 25 U/mg
ICN15084401 1 mg ≈40 U/mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15084405 Supplier MP Biomedicals™ Supplier No. 0215084405
Add to Cart
Add to Cart

Diaphorase is used in a study to assess the protein-protein interactions in assembly of lipoic acid on 2-oxoacid dehydrogenases of aerobic metabolism

  • CAS Number: 9001-18-7
  • EC Number: 232-587-6
  • MDL Number: MFCD00130947
  • Physical Appearance: Pale yellow, freeze dried powder
  • Specific Activity: Approx. 10U/mg protein
  • Format: Freeze-dried powder in presence of sucrose
  • Soluble in distilled water or dilute buffer

TRUSTED_SUSTAINABILITY

Specifications

Concentration 25 U/mg
Color Pale Yellow
For Use With (Application) Study the assess the Protein-Protein interactions in assembly of Lipoic Acid on the 2-oxoacid Dehydrogenases of aerobic metabolism
Content And Storage 0°C
Source Procine heart
Quantity 5 mg
Product Type Diaphorase
Form Lyophilized Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.