Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Dispase

Catalog No. p-3563420 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 U
5 U
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19502205 5 U
ICN19502225 25 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19502205 Supplier MP Biomedicals™ Supplier No. 0219502205
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

From Bacillus polymyxa

Microbial metalloenzyme used in tissue dissociation (primary cell cultures) and subcultivation (secondary cell cultures). Extremely stable. Activity: 6U/mg. Unit Definition: One unit is the amount of enzyme which liberates 1.0μmole of Tyr per minute at 37°C, pH 7.5. MW: 35.9kDa. Lyophilized.

Ref.: Akimoto, Y., et al., J. Histochem. Cytochem., 40: 1129 (1992).

TRUSTED_SUSTAINABILITY

Specifications

Color Tan
pH 6 to 8.5
For Use With (Application) Protein sequencing
Content And Storage 4°C
Source Bacillus polymyxa
Quantity 5 U
Product Type Dispase
Molecular Weight (g/mol) 35.9
Inhibitors EDTA, EGTA, Hg2+, other heavy Metals
Form Lyophilized Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.