Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ DOB 2% Galactose (glucose-free), Powdered Media

Catalog No. MP114025522 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
454 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP114025522 454 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP114025522 Supplier MP Biomedicals™ Supplier No. 114025522
Add to Cart
Add to Cart

Can be used to create SD or SDA by combining with the appropriate amino acid mixture

Formulation

  • Use 26.7g powder per liter
  • Contents/liter: 1.7g YNB, 5g Ammonium Sulfate, 20g Galactose

Yeast cell culture, Gene expression

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Molecular biology Certified™
Content And Storage 15°C to 30°C
Form Powder
Product Type Yeast Media
Formulation YNB 1.7g/L, Ammonium Sulfate 5g/L, Galactose 20g/L
For Use With (Application) For Saccharomyces cerevisiae growth medium and Yeast cell culture, gene expression
Quantity 454 g
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.