Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ DOBA Medium, Powdered Media

Catalog No. MP114026022 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
454 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP114026022 454 g
MP114026032 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP114026022 Supplier MP Biomedicals™ Supplier No. 114026022
Add to Cart
Add to Cart

Can be used to create SD or SDA by combining with the appropriate amino acid mixture

Formulation

  • Use 43.7g powder liter
  • Contents/liter: DOB, 17g Agar-Y

Applications:

Yeast cell culture, Gene expression, Transcription regulation

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Molecular biology Certified™
Content And Storage 15°C to 30°C
Form Powder
Grade Molecular biology
Product Type Yeast Media
Formulation Nitrogen Sources : Ammonium Sulfate 5g/L, Carbon Source: Dextrose 20g/L, Vitamins : Biotin 0.002mg/L, Calcium Pantothenate 0.4mg/L, Folic Acid 0.002mg/L, Inositol 2mg/L, Niacin 0.4mg/L, P-Aminobenzoic Acid 0.2mg/L, Pyridoxine Hydrochloride 0.4mg/L, Riboflavin 0.2mg/L, Thiamine Hydrochloride 0.4mg/L, Trace Elements: Boric Acid 0.5mg/L, Copper Sulfate 0.04mg/L, Potassium Iodide 0.1mg/L, Ferric Chloride 0.2mg/L, Manganese Sulfate 0.4mg/L, Sodium Molybdate 0.2mg/L, Zinc Sulfate 0.4mg/L, Salts: Potassium Phosphate, Monobasic 1000mg/L, Magnesium Sulfate 500mg/L, Sodium Chloride 100mg/L, Calcium Chloride 100mg/L, Agar 17g/L
For Use With (Application) For Yeast cell culture, gene expression, transcription regulation
Quantity 454 g
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.