Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Elastase 2X Crystallized, from Porcine Pancreas

Catalog No. ICN10061780 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10061780 100 mg
ICN10061725 25 mg
ICN10061710 10 mg
ICN10061750 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN10061780 Supplier MP Biomedicals™ Supplier No. 0210061780
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Activity 50 u/mg protein, Porcine Pancreas, Milky Solution

Elastase was used to study human emphysema by induction of emphysema in rodents. It has been used in a study to investigate the design, synthesis and evaluation of biomimetic affinity ligands for elastases. Elastase from porcine pancreas has also been used in a study to investigate the purification and partial characterization of the pancreatic proteolytic enzymes trypsin, chymotrypsin, and elastase. It hydrolyzes peptide bonds, especially those adjacent to neutral amino acids.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X
Color Milky
For Use With (Application) Investigate the purification and partial characterization of the pancreatic proteolytic Enzymes
Content And Storage 4°C
Source Porcine pancreas
Quantity 100 mg
Product Type Elastase
Molecular Weight (g/mol) ≈25000
Inhibitors DFP, Elastinal, alpha-2-macroglobulin
Form Solution
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.