Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Elastin from Bovine Neck Ligament

Catalog No. ICN10163601 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10163601 1 g
ICN10163605 5 g
ICN10163625 25 g
ICN10163610 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN10163601 Supplier MP Biomedicals™ Supplier No. 0210163601
Add to Cart
Add to Cart

Elastin is a structural protein which is highly stable and hemocompatible in nature

Elastin is responsible for the elasticity of many tissues. It is especially important in blood vessels, where it comprises about 50% of the dry tissue weight.

  • CAS Number: 9007-58-3
  • EC Number: 232-701-4
  • MDL Number: MFCD00130999
  • Format: off white powder

Used in construction of biomaterials

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) COPD Studies
Name Elastin
Quantity 1 g
Source Bovine
Storage Requirements 4°C
Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.