Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Factor IXa

Catalog No. ICN19419480 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19419480 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19419480 Supplier MP Biomedicals™ Supplier No. 0219419480

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

From human plasma

Purified.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Involved in the activation of the Zymogen, factor X, to form the Enzyme, factor Xa
Content And Storage -20°C
Source Human Plasma
Quantity 100 μg
Product Type Factor IXa
Molecular Weight (g/mol) 45
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.