Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Ficin

Catalog No. ICN19876180 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19876180 100 g
ICN19876105 5 g
ICN19876125 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN19876180 Supplier MP Biomedicals™ Supplier No. 0219876180
Add to Cart
Add to Cart

From fig tree

Activity: 300-500 milk clotting units/mg. Hygroscopic.

TRUSTED_SUSTAINABILITY

Specifications

Color White
pH 5 to 7.5
For Use With (Application) Cleavage of Mouse IgG
Content And Storage 4°C
Source Fig tree Ficus glabrata
Quantity 100 g
Product Type Ficin
Molecular Weight (g/mol) ≈25
Inhibitors Iodoacetamide, Iodoacetic Acid, N-ethylmaleimide, Mercuric Chloride, DFP (Diisopropyl Fluorophosphate), TLCK (Na-p-Tosyl-lysine Chloromethyl Ketone), TPCK (N-Tosyl-L-phenylalanine Chloromethyl Ketone)
Form Powder
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.