Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Follicle Stimulating Hormone

Catalog No. ICN10172750 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10172750 50 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10172750 Supplier MP Biomedicals™ Supplier No. 0210172750
Only null left
Add to Cart
Add to Cart

A two-chain glycoprotein gonadotropic hormone

Follicle stimulating hormone induces maturation of the Graafian follicles of the ovary and promotes the development of germinal cells in males. FSH regulates Sertoli cells by acting on G-protein-linked cell surface FSH receptors. It activates cytosolic soluble protein tyrosine kinase (CyPTk).

TRUSTED_SUSTAINABILITY

Specifications

Name Follicle Stimulating Hormone
Quantity 50 U
Source Porcine Pituitary
Storage Requirements Store at 0°C
Form Crystalline
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.