Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Freund's Incomplete Adjuvant

Catalog No. ICN55829 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN55829 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN55829 Supplier MP Biomedicals™ Supplier No. 0855829
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used with subsequent boosts to increase antibody titer following initial animal injection with Freund's Complete Adjuvant

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
For Use With (Application) Used with subsequent boosts to increase antibody titer following initial animal injection with Freund's complete adjuvant, immunoassays, as boost injections
Formulation Paraffin oil 80 to 90%, Arlacel A 10 to 20%
Quantity 25 mL
Product Type Freund's Incomplete Adjuvant
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.