Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ β- Galactosidase from Escherichia coli

Catalog No. icn10493901 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 kU
10 kU
5 kU
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10493901 1 kU
ICN10493905 5 kU
ICN10493910 10 kU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN10493901 Supplier MP Biomedicals™ Supplier No. 0210493901
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

β-galactosidase cleaves lactose into its monosaccharide components, glucose and galactose

  • CAS Number: 9031-11-2
  • EC Number: 232-864-1
  • Format: Pale gray fuffy powder
  • Specific activity: ≥50U/mg
  • Isoelectric point: 4.61
  • Optimum pH: 7 to 7.5

Used for structural investigation of carbohydrate, determination of lactose (foodstuff analysis) and as an enzyme label for enzyme immunoassay.

TRUSTED_SUSTAINABILITY

Specifications

Concentration ≥50 U/mg
Color Pale Gray
pH 7 to 7.5
For Use With (Application) Structural investigation of Carbohydrate, the determination of Lactose (food stuff analysis) and as an Enzyme label for Enzyme immunoassay
Content And Storage 4°C
Source E. coli
Quantity 1 kU
Product Type beta-Galactosidase
Molecular Weight (g/mol) 540000
Inhibitors p-Chloromercuribenzoate, lodoacetamide, Heavy Metal Ions (Zn++, Fe++, Zn++, Cd++, Cu++, Pb++, Ag+, Hg++), Ionic Detergents SDS, DAC
Form Lyophilized Powder
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.