Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
- CAS Number: 9031-11-2
- EC Number: 232-864-1
- Format: Pale gray fuffy powder
- Specific activity: ≥50U/mg
- Isoelectric point: 4.61
- Optimum pH: 7 to 7.5
Used for structural investigation of carbohydrate, determination of lactose (foodstuff analysis) and as an enzyme label for enzyme immunoassay.
Specifications
Specifications
| Concentration | ≥50 U/mg |
| Color | Pale Gray |
| pH | 7 to 7.5 |
| For Use With (Application) | Structural investigation of Carbohydrate, the determination of Lactose (food stuff analysis) and as an Enzyme label for Enzyme immunoassay |
| Content And Storage | 4°C |
| Source | E. coli |
| Quantity | 1 kU |
| Product Type | beta-Galactosidase |
| Molecular Weight (g/mol) | 540000 |
| Inhibitors | p-Chloromercuribenzoate, lodoacetamide, Heavy Metal Ions (Zn++, Fe++, Zn++, Cd++, Cu++, Pb++, Ag+, Hg++), Ionic Detergents SDS, DAC |
| Show More |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?