Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Glucagon, Human

Catalog No. ICN15287901 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
0.001g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15287901 0.001g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15287901 Supplier MP Biomedicals™ Supplier No. 0215287901
Add to Cart
Add to Cart

  • Hyperglycemic-glycogenolytic factor found in the pancreas, an antihypoglycemic agent

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1mg/mL
For Use With (Application) Antihypoglycemic Agent
Molecular Weight (g/mol) 3482.81
Name Glucagon (29 Amino Acids) from Human
Quantity 0.001g
Source Human
Storage Requirements -20°C
Form Solid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.