Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Glucose Oxidase

Catalog No. icn19519683 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 kU
250 kU
50 kU
500 kU
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19519683 250 kU
ICN19519610 10 kU
ICN19519650 50 kU
ICN19519690 500 kU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN19519683 Supplier MP Biomedicals™ Supplier No. 0219519683
Add to Cart
Add to Cart

From Aspergillus niger

Type X. Activity: >100U/mg. Unit Definition: one unit oxidizes 1.0μmol of β-D-glucose to D-gluconic acid and H2O2 per minute at 35°C, pH 5.1.

TRUSTED_SUSTAINABILITY

Specifications

Concentration >100 U/mg
Color Yellow
pH 4 to 7
For Use With (Application) Glucose Oxidase can be utilized in the enzymatic determination of D-glucose in solution
Content And Storage -20°C
Source Aspergillus niger
Quantity 250 kU
Product Type Glucose Oxidase
Molecular Weight (g/mol) ≈160000
Inhibitors Ag+, Hg2+, Cu2+ , 4-chloromercuribenzoate, 50% D-arabinose
Form Lyophilized Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.