Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ GS96 Media, Powdered

Catalog No. MP113101132 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
227 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP113101132 1000 g
MP113101112 227 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP113101132 Supplier MP Biomedicals™ Supplier No. 113101132
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High yields of plasmid DNA in 96-well and 384-well plates

GS96 Broth is formulated to grow host cells to the highest density possible without aeration, and to produce high yields of plasmid DNA from those cultures. Perfect for growing bacterial cells in 96-well and 384-well plates, GS96 media can be used in any situation in which aeration of the culture is difficult or absent. GS96 broth is used at the front end of high-throughput automated procedures, and growth can be increased even further by the addition of 0.05% glycerol.

  • Formulated to grow host cells to the highest density possible without aeration, and to produce high yields of plasmid DNA from those cultures
  • Perfect for growing bacterial cells in 96- and 384-well plates
  • Can be used in any situation in which aeration of the culture is difficult or absent
  • Used at the front end of high-throughput automated procedures
  • Growth can be increased even further by the addition of 0.05% glycerol
Formulation
  • Contents/L: proprietary
  • Use 50g/L

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15°C to 30°C
Form Powder
Grade Molecular biology
Product Type Microbiology Media
Formulation Yeast extract, Peptones, Vitamins, Trace Elements, Carbon and Nitrogen Sources, Small quantities of biological Buffers have been added for proper pH adjustment
For Use With (Application) To grow host cells to the highest density possible without aeration and to produce high yields of plasmid DNA from those cultures
Quantity 1000 g
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.