Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Hemocyanin

Catalog No. ICN15123383 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15123383 250 mg
ICN15123350 50 mg
ICN15123380 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15123383 Supplier MP Biomedicals™ Supplier No. 0215123383
Add to Cart
Add to Cart

Derived from Megathura crenulata

Hemocyanins are copper-containing oxygen transport proteins found in the hemolymph of many invertebrates. It is dissolved in the hemolymph; not found in blood cells. They are divided into 2 main groups, arthropodan (such as horseshoe crab) and molluscan (such as keyhole limpet). These have structurally similar oxygen-binding sites, which are similar to the oxygen-binding sites of tyrosinases, but their quaternary structures are arranged differently.

  • Format: Gray lyophilized powder

Immunology

TRUSTED_SUSTAINABILITY

Specifications

Formulation 5mM BES Buffer, pH 6.5 and 10mM MgSO4
Molecular Weight (g/mol) 3000000 to 7500000
Name Hemocyanin
Quantity 250 mg
Source Keyhole Limpet
Storage Requirements 4°C
Form Lyophilized
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.