Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Prostaglandin E2, γ-irradiated, ≥99%

Catalog No. ICN19457601 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19457601 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19457601 Supplier MP Biomedicals™ Supplier No. 0219457601
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Prostaglandin E2 is used in cell culture applications for the study of prostaglandin regulated cell signaling and gene regulation.

Prostaglandin E2 (PGE2) is an arachidonic acid derivative and important inflammatory mediator in immune responses. Its activity has been shown to influence inflammation, fertility and parturition, gastric mucosal integrity, and immune modulation. PGE2 is used in cell culture applications to study prostaglandin regulated cell signaling and gene regulation.

TRUSTED_SUSTAINABILITY

Specifications

CAS 363-24-6
Content And Storage -20°C
Form Solid
Sterility Sterile
Sterilization Method Gamma-irradiated
Purity or Quality Grade ≥99%
Quantity 1 mg
Source Synthetic

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.